Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rrs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200062.1 | NP_956356.1 | 1 | 351 | 100 | |
GenBank mRNA | BC055615.1 | - | 1 | 351 | 100 | |
BC164156.1 | - | 1 | 376 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |