Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cfap298 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200088.1 | NP_956382.1 | 7 - 8 | 859 | 76 | |
GenBank mRNA | AY648836.1 | - | 7 - 8 | 840 | 76 | |
BC053230.1 | - | 7 - 8 | 859 | 76 | ||
BC154333.1 | - | 6 - 7 | 880 | 76 | ||
BC171713.1 | - | 7 - 8 | 844 | 76 | ||
BC171715.1 | - | 7 - 8 | 844 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |