Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gbgt1l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200125.1 | NP_956419.1 | 2 - 3 | 138 | 83 | |
XM_005165128.4 | XP_005165185.1 | 2 - 3 | 187 | 83 | ||
XM_005165129.4 | XP_005165186.1 | 2 - 3 | 213 | 83 | ||
GenBank mRNA | BC055647.1 | - | 2 - 3 | 138 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |