Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stard14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200220.1 | NP_956514.1 | 3 - 4 | 580 | 82 | |
XM_005173231.3 | XP_005173288.1 | 4 - 5 | 552 | 82 | ||
GenBank mRNA | BC046038.1 | - | 3 - 4 | 580 | 82 | |
BC063977.1 | - | 3 - 4 | 492 | 82 | ||
BC171583.1 | - | 3 - 4 | 508 | 82 | ||
BC171587.1 | - | 3 - 4 | 508 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |