Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sh3bgrl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200278.2 | NP_956572.1 | 2 - 3 | 317 | 105 | |
GenBank mRNA | BC049323.1 | - | 2 - 3 | 105 | ||
EH440040.1 | - | 2 - 3 | 349 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |