Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxo43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200431.2 | NP_956725.2 | 7 - 8 | 1606 | 95 | |
XM_021466645.1 | XP_021322320.1 | 7 - 8 | 1618 | 95 | ||
GenBank mRNA | BC097058.1 | - | 7 - 8 | 1610 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |