Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pex13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200645.1 | NP_956939.1 | 1 - 2 | 209 | 82 | |
GenBank mRNA | BC044529.1 | - | 3 - 4 | 2697 | 82 | |
BC057464.1 | - | 1 - 2 | 209 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |