Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btf3l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200694.1 | NP_956988.1 | 2 - 3 | 121 | 59 | |
XM_009305886.2 | XP_009304161.1 | 3 - 4 | 211 | 59 | ||
XM_009305887.3 | XP_009304162.1 | 2 - 3 | 138 | 59 | ||
XM_021479564.1 | XP_021335239.1 | 3 - 4 | 211 | 59 | ||
GenBank mRNA | BC059432.1 | - | 2 - 3 | 121 | 59 | |
BC150462.1 | - | 3 - 4 | 173 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |