Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndufb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200730.1 | NP_957024.1 | 2 - 3 | 369 | 99 | |
GenBank mRNA | BC059507.1 | - | 2 - 3 | 369 | 99 | |
BC071489.1 | - | 2 - 3 | 367 | 99 | ||
BC154202.1 | - | 2 - 3 | 372 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |