Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arl5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200846.1 | NP_957140.1 | 3 - 4 | 466 | 65 | |
GenBank mRNA | BC062281.1 | - | 3 - 4 | 466 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |