Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200897.2 | NP_957191.1 | 3 - 4 | 374 | 61 | |
GenBank mRNA | BC064303.1 | - | 3 - 4 | 366 | 61 | |
EH446923.1 | - | 3 - 4 | 385 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |