Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: timm17b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113593.1 | NP_001107065.1 | 5 - 6 | 453 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Danio rerio Probe spans exons |