Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cacna1ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114548.1 | NP_001108020.1 | 40 - 41 | 5445 | 81 | |
XM_009301497.3 | XP_009299772.1 | 44 - 45 | 6279 | 81 | ||
XM_009301500.3 | XP_009299775.1 | 42 - 43 | 5610 | 81 | ||
XM_009301501.3 | XP_009299776.1 | 43 - 44 | 5580 | 81 | ||
XM_009301504.3 | XP_009299779.1 | 42 - 43 | 5610 | 81 | ||
XM_009301505.3 | XP_009299780.1 | 40 - 41 | 6198 | 81 | ||
XM_017356095.2 | XP_017211584.1 | 44 - 45 | 5616 | 81 | ||
XM_017356096.2 | XP_017211585.1 | 43 - 44 | 5601 | 81 | ||
XM_017356097.2 | XP_017211586.1 | 42 - 43 | 6210 | 81 | ||
XM_021476173.1 | XP_021331848.1 | 44 - 45 | 5643 | 81 | ||
XM_021476174.1 | XP_021331849.1 | 43 - 44 | 5637 | 81 | ||
XM_021476175.1 | XP_021331850.1 | 43 - 44 | 5631 | 81 | ||
XM_021476176.1 | XP_021331851.1 | 43 - 44 | 5622 | 81 | ||
XM_021476178.1 | XP_021331853.1 | 41 - 42 | 5604 | 81 | ||
XM_021476179.1 | XP_021331854.1 | 41 - 42 | 5604 | 81 | ||
XM_021476180.1 | XP_021331855.1 | 44 - 45 | 5643 | 81 | ||
XM_021476181.1 | XP_021331856.1 | 41 - 42 | 6183 | 81 | ||
XM_021476182.1 | XP_021331857.1 | 40 - 41 | 6177 | 81 | ||
GenBank mRNA | KC192784.1 | - | 42 - 43 | 5610 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |