Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zeb2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114551.2 | NP_001108023.2 | 8 - 9 | 3023 | 57 | |
NM_001135104.2 | NP_001128576.2 | 8 - 9 | 3023 | 57 | ||
XM_005167523.4 | XP_005167580.4 | 7 - 8 | 3172 | 57 | ||
XM_021478582.1 | XP_021334257.1 | 8 - 9 | 3320 | 57 | ||
XM_021478583.1 | XP_021334258.1 | 7 - 8 | 3266 | 57 | ||
GenBank mRNA | BC168514.1 | - | 8 - 9 | 3003 | 57 | |
EU379558.1 | - | 7 - 8 | 2834 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |