Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmem26b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114592.1 | NP_001108064.1 | 5 - 6 | 711 | 93 | |
GenBank mRNA | BC154616.1 | - | 5 - 6 | 711 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |