Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vwc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114628.1 | NP_001108100.1 | 2 - 3 | 775 | 54 | |
XM_005156699.4 | XP_005156756.1 | 3 - 4 | 1282 | 54 | ||
GenBank mRNA | AB292672.1 | - | 2 - 3 | 775 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Danio rerio Probe spans exons |