Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyth4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114903.2 | NP_001108375.2 | 7 - 8 | 573 | 73 | |
XM_009302077.2 | XP_009300352.1 | 4 - 5 | 470 | 73 | ||
GenBank mRNA | BC154839.1 | - | 7 - 8 | 553 | 73 | |
EH453546.1 | - | 7 - 8 | 604 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |