Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arid1aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021466949.1 | XP_021322624.1 | 6 - 7 | 2965 | 71 | |
XM_021466950.1 | XP_021322625.1 | 6 - 7 | 2964 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |