Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btbd6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114925.1 | NP_001108397.1 | 2 - 3 | 582 | 79 | |
XM_005170161.4 | XP_005170218.1 | 3 - 4 | 489 | 79 | ||
GenBank mRNA | BC155622.1 | - | 2 - 3 | 582 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |