Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tgfbr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115059.1 | NP_001108531.1 | 2 - 3 | 407 | 88 | |
GenBank mRNA | EF462418.1 | - | 2 - 3 | 407 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |