Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: edar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115064.2 | NP_001108536.2 | 11 - 12 | 1184 | 106 | |
GenBank mRNA | EF137867.1 | - | 11 - 12 | 1181 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Danio rerio Probe spans exons |