Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdc25b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115095.2 | NP_001108567.1 | 13 - 14 | 1617 | 107 | |
GenBank mRNA | CT708651.2 | - | 1 - 2 | 84 | 107 | |
EU921896.1 | - | 13 - 14 | 1595 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons |