Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdc25b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115095.2 | NP_001108567.1 | 2 - 3 | 605 | 72 | |
GenBank mRNA | EH500454.1 | - | 2 - 3 | 618 | 72 | |
EU921896.1 | - | 2 - 3 | 586 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Danio rerio Probe spans exons |