Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcdh18b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115099.1 | NP_001108571.1 | 1 - 2 | 2581 | 90 | |
GenBank mRNA | AB297803.1 | - | 1 - 2 | 2581 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Danio rerio Probe spans exons |