Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gapdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115114.1 | NP_001108586.1 | 9 - 10 | 817 | 79 | |
GenBank mRNA | BC083506.1 | - | 9 - 10 | 817 | 79 | |
BC095386.1 | - | 9 - 10 | 817 | 79 | ||
BC115131.1 | - | 9 - 10 | 821 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |