Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139480.1 | NP_001132952.1 | 2 - 3 | 842 | 85 | |
XM_005173662.4 | XP_005173719.1 | 2 - 3 | 856 | 85 | ||
GenBank mRNA | BC129216.1 | - | 2 - 3 | 811 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |