Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139480.1 | NP_001132952.1 | 3 - 4 | 960 | 123 | |
GenBank mRNA | BC129216.1 | - | 3 - 4 | 929 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |