Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpp6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115122.1 | NP_001108594.1 | 22 - 23 | 2200 | 69 | |
XM_005163289.3 | XP_005163346.1 | 17 - 18 | 2168 | 69 | ||
XM_005163290.4 | XP_005163347.1 | 17 - 18 | 1643 | 69 | ||
XM_005163291.4 | XP_005163348.1 | 17 - 18 | 1611 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |