Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115133.1 | NP_001108605.1 | 1 - 2 | 205 | 114 | |
GenBank mRNA | BC056715.1 | - | 1 - 2 | 176 | 114 | |
DR722002.1 | - | 1 - 2 | 224 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |