Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115133.1 | NP_001108605.1 | 7 - 8 | 707 | 134 | |
GenBank mRNA | BC056715.1 | - | 7 - 8 | 678 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |