Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prom1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115143.2 | NP_001108615.2 | 4 - 5 | 709 | 57 | |
XM_005157347.3 | XP_005157404.1 | 4 - 5 | 834 | 57 | ||
XM_005157349.3 | XP_005157406.1 | 5 - 6 | 862 | 57 | ||
XM_005157351.3 | XP_005157408.1 | 5 - 6 | 862 | 57 | ||
XM_009291267.3 | XP_009289542.1 | 5 - 6 | 964 | 57 | ||
XM_009291269.3 | XP_009289544.1 | 4 - 5 | 834 | 57 | ||
XM_021480968.1 | XP_021336643.1 | 5 - 6 | 965 | 57 | ||
XM_021480969.1 | XP_021336644.1 | 5 - 6 | 965 | 57 | ||
XM_021480970.1 | XP_021336645.1 | 4 - 5 | 938 | 57 | ||
XM_021480971.1 | XP_021336646.1 | 5 - 6 | 964 | 57 | ||
XM_021480972.1 | XP_021336647.1 | 5 - 6 | 964 | 57 | ||
XM_021480973.1 | XP_021336648.1 | 4 - 5 | 835 | 57 | ||
XM_021480974.1 | XP_021336649.1 | 5 - 6 | 965 | 57 | ||
XM_021480975.1 | XP_021336650.1 | 5 - 6 | 964 | 57 | ||
GenBank mRNA | AF160970.1 | - | 4 - 5 | 834 | 57 | |
BC163446.1 | - | 4 - 5 | 709 | 57 | ||
HQ386793.1 | - | 5 - 6 | 866 | 57 | ||
HQ386794.1 | - | 5 - 6 | 869 | 57 | ||
HQ386795.1 | - | 4 - 5 | 834 | 57 | ||
HQ386796.1 | - | 4 - 5 | 846 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |