Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptbp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122654.1 | NP_001116126.1 | 15 - 16 | 1671 | 80 | |
XM_005155938.4 | XP_005155995.1 | 14 - 15 | 1600 | 80 | ||
XM_009305880.2 | XP_009304155.1 | 15 - 16 | 1620 | 80 | ||
GenBank mRNA | BC091854.1 | - | 15 - 16 | 1633 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |