Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040606.1 | 1 - 2 | 83 | 65 | ||
GenBank mRNA | EU520304.1 | - | 1 - 2 | 68 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |