Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mxg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122971.1 | NP_001116443.1 | 9 - 10 | 1261 | 108 | |
XM_009297890.2 | XP_009296165.1 | 9 - 10 | 1484 | 108 | ||
XM_021470456.1 | XP_021326131.1 | 7 - 8 | 988 | 108 | ||
GenBank mRNA | AJ544829.2 | - | 9 - 10 | 1261 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons |