Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131036.2 | NP_571111.2 | 2 - 3 | 549 | 92 | |
XM_005171264.4 | XP_005171321.1 | 2 - 3 | 544 | 92 | ||
XM_005171265.3 | XP_005171322.1 | 2 - 3 | 544 | 92 | ||
XM_009298633.3 | XP_009296908.1 | 2 - 3 | 544 | 92 | ||
GenBank mRNA | BC071284.1 | - | 2 - 3 | 512 | 92 | |
BC095389.1 | - | 2 - 3 | 471 | 92 | ||
KM655642.1 | - | 1 - 2 | 383 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |