Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srsf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199574.1 | NP_955868.1 | 3 - 4 | 415 | 96 | |
GenBank mRNA | BC046895.1 | - | 3 - 4 | 415 | 96 | |
BC164410.1 | - | 3 - 4 | 408 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |