Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: derl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200903.2 | NP_957197.2 | 3 - 4 | 445 | 90 | |
GenBank mRNA | BC064666.1 | - | 3 - 4 | 444 | 90 | |
CK026866.1 | - | 3 - 4 | 371 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |