Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: phkg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200962.1 | NP_957256.1 | 7 - 8 | 847 | 89 | |
XM_009306599.3 | XP_009304874.1 | 7 - 8 | 860 | 89 | ||
GenBank mRNA | BC047191.1 | - | 7 - 8 | 847 | 89 | |
BC075780.1 | - | 7 - 8 | 837 | 89 | ||
BC152636.1 | - | 7 - 8 | 807 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |