Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nck2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200997.1 | NP_957291.1 | 2 - 3 | 325 | 84 | |
XM_009304471.3 | XP_009302746.1 | 3 - 4 | 589 | 84 | ||
XM_021478633.1 | XP_021334308.1 | 2 - 3 | 2067 | 84 | ||
GenBank mRNA | BC046872.1 | - | 2 - 3 | 325 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |