Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prkacba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201023.1 | NP_957317.1 | 9 - 10 | 1153 | 78 | |
GenBank mRNA | BC053227.1 | - | 9 - 10 | 1153 | 78 | |
BC165279.1 | - | 9 - 10 | 1087 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |