Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: coro1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201114.1 | NP_957408.1 | 4 - 5 | 567 | 61 | |
GenBank mRNA | AY330221.1 | - | 4 - 5 | 547 | 61 | |
BC055237.1 | - | 4 - 5 | 567 | 61 | ||
BC161664.1 | - | 4 - 5 | 535 | 61 | ||
BC171389.1 | - | 4 - 5 | 519 | 61 | ||
BC171393.1 | - | 4 - 5 | 519 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |