Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nedd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201321.2 | NP_958478.2 | 2 - 3 | 213 | 91 | |
GenBank mRNA | BC055645.1 | - | 2 - 3 | 210 | 91 | |
EH443336.1 | - | 2 - 3 | 239 | 91 | ||
EH507246.1 | - | 2 - 3 | 216 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |