Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201329.1 | NP_958486.1 | 1 - 2 | 520 | 96 | |
GenBank mRNA | BC053195.1 | - | 1 - 2 | 520 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |