Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aldh9a1a.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201471.1 | NP_958879.1 | 9 - 10 | 1249 | 88 | |
XM_005166774.3 | XP_005166831.1 | 9 - 10 | 1354 | 88 | ||
GenBank mRNA | BC045932.1 | - | 9 - 10 | 1249 | 88 | |
BC066668.1 | - | 9 - 10 | 1242 | 88 | ||
BC165753.1 | - | 8 - 9 | 1212 | 88 | ||
BC166038.1 | - | 9 - 10 | 1251 | 88 | ||
BC167472.1 | - | 9 - 10 | 1251 | 88 |
Gene Symbol: aldh9a1a.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126480.1 | NP_001119952.1 | 8 - 9 | 1201 | 88 | |
XM_021478161.1 | XP_021333836.1 | 8 - 9 | 1196 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |