Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mmp13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290479.1 | NP_001277408.1 | 7 - 8 | 1107 | 89 | |
GenBank mRNA | BC049472.1 | - | 7 - 8 | 1028 | 89 | |
BC065591.1 | - | 7 - 8 | 1078 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |