Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: micall2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001327787.1 | NP_001314716.1 | 4 - 5 | 892 | 68 | |
NR_137174.1 | 4 - 5 | 892 | 68 | |||
XM_005164198.4 | XP_005164255.1 | 4 - 5 | 892 | 68 | ||
GenBank mRNA | BC052767.1 | - | 4 - 5 | 794 | 68 | |
HM151009.1 | - | 4 - 5 | 654 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |