Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: micall2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_137174.1 | 8 - 9 | 1821 | 89 | ||
GenBank mRNA | BC052767.1 | - | 8 - 9 | 1723 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |