Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rabl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205733.1 | NP_991296.1 | 2 - 3 | 138 | 109 | |
GenBank mRNA | AY391440.1 | - | 2 - 3 | 138 | 109 | |
BC078191.1 | - | 2 - 3 | 177 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |