Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttn.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021479071.1 | XP_021334746.1 | 65 - 66 | 14760 | 71 | |
XM_021479072.1 | XP_021334747.1 | 23 - 24 | 7159 | 71 | ||
GenBank mRNA | CT664813.2 | - | 1 - 2 | 402 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Danio rerio Probe spans exons |