Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ufm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212627.1 | NP_997792.1 | 5 - 6 | 222 | 111 | |
GenBank mRNA | AY394956.1 | - | 5 - 6 | 222 | 111 | |
BC044145.1 | - | 5 - 6 | 263 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |